Learn More
Whatman™ Grade P81 Ion Exchange Cellulose Chromatography Paper
Manufactured from pure cellulose and ion-exchange cellulose fibers to permit highly efficient, rapid separations of charged organic or inorganic substances
Manufacturer:  Whatman™ 3698325
 View more versions of this product
Catalog No. 05-717-2B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
2.5cm
For separation of biogenic Amines, antibiotics, Histamines, and certain Metallic CatIons at very low concentrations
P81
Cellulose, Ion Exchange Cellulose Fiber
0.23mm
Circle
18μeq/cm2
125/30mm/min.