Learn More
Whatman™ Harris Micro Punch
Sharp cutting edge provides quick and easy sampling from FTA™ and CloneSaver™ Cards with precise punching and ejection, uniform sampling, and no carryover between punches
Manufacturer:  Whatman™ WB100061
 View more versions of this product
Catalog No. 09-301-261
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Polished-steel tips are precision tooled, heat treated, rust resistant, and case hardened
  • Self-healing inert cutting mat ensures clean sample cuts and extends the life of the punch
  • 2.0mm size is recommended for CloneSaver Cards
  • Replacement tips and cutting mats are available separately