Available on GSA/VA Contract for Federal Government customers only. Whatman™ HEPA-Vent Filter
For filtration of air and gas
$109.98
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
09-744-79
Available on GSA/VA Contract for Federal Government customers only. 
Whatman
6723-5000
Filter area: 16cm2; Inlet/Outlet: 1/4 to 3/8 in. stepped barb
Pack of 10 for $109.98
Check Availability
Add to Cart
Descriptions & Specifications
Related Products