Learn More
GE Healthcare Whatman™ Klari-Flex™ Bottle-top Filtration System
Provide vacuum filtration of aqueous solutions such as cell culture media, biological fluids, and buffers
Manufacturer:  GE Healthcare 65142502
 View more versions of this product
Catalog No. 09-744-301
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Drop-Connect technology ensures fail-safe vacuum connection and unit placement in base.
  • Weighted pedestal base and wide, low-profile funnel/bottle filtration units are stable and tip resistant, minimizing the risk of spills and contamination.
  • Graduation marks face front for readability.
  • Large bottle cap accommodates labeling information.
  • Ergonomically designed bottle and bottle cap provide improved handling.
  • Membrane material is PES.