Learn More
Cytiva Whatman™ Lens Cleaning Tissue PROMO
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Manufacturer:  Cytiva 29648081
Catalog No. 08-777-100LR1
Add to cart
Description
Description
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Soft textured strong tissue leaves no fibers. Protects lenses and other optical surfaces made from glass, quartz, or plastic from scratches.
  • Chemically pure, free of silicone and other additives
  • High absorbency leads to increased safety upon removal of surface moisture and grease
  • Thickness 0.035 to 0.040mm
  • Grade 105 sheets
Specifications
Specifications
150 x 100 mm
25 wallets of 25 sheets
Safe removal of surface moisture and grease.
Won't damage lenses or optical surfaces, leaves no fibers
Lens wipe