Learn More
GE Healthcare™ Whatman™ Microplate Heat Seals
Manufacturer:  GE Healthcare™ 77040003
Catalog No. 09-003-41
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Controls humidity and reduces evaporation of samples.
  • Prevents spills and contamination.
  • Cold seals are self-sticking with inert adhesive.
  • Heat seals are available in a clear polyester or aluminum foil.
  • Heat seals are for polypropylene microplates only and are applied with heat and pressure.
Description & Specifications
Specifications
Polyethylene terephthalate (PET)
Used to control humidity and reduce evaporation of samples
100Pack
0.13mm
Heat, pressure-applied
Clear
Microplate Seal