Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Mini-UniPrep™ Amber Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.

Produced with the same colorant used in pharmaceutical containers. Whatman™ Mini-UniPrep™ Amber Syringeless Filters provide an easier way to remove particulates from HPLC/UHPLC samples in one-third the time as compared to traditional syringe filters.

$442.00 - $522.00

Specifications

Color Amber
Material Septa: polytetrafluoroethylene (PTFE) coated silicone rubber
Housing Material Polypropylene (PP)
Pressure Rating 0.6 bar / 8 psi (to compress)
Volume (Metric) 0.4mL
View More Specs

Products 10
Catalog Number Mfr. No. Membrane Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Membrane Pore Size No. per Pack Price Quantity & Availability  
09-923-248
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APEAQU
Polyvinylidene Difluoride (PVDF) 0.2μm 100/Pk.
Pack of 100 for $522.00
Add to cart
 
09-923-254
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APUDPP
Polypropylene Depth Filter (Depth PP) 0.45μm 100/Pk.
Pack of 100 for $522.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-923-256
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APUORG
Polytetrafluoroethylene (PTFE) 0.45μm 100/Pk.
Pack of 100 for $442.00
Add to cart
 
09-923-258
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APUPP
Polypropylene (PP) 0.45μm 100/Pk.
Pack of 100 for $522.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-927-631
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APUGMF
Glass Microfiber (GMF) 0.45μm 100/Pk.
Pack of 100 for $522.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-927-632
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APEPES
Polyethersulfone (PES) 0.2μm 100/Pk.
Pack of 100 for $522.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-927-815
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APENYL
Nylon (NYL) 0.2μm 100/Pk.
Pack of 100 for $522.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-924-584 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APEORG
Polytetrafluoroethylene (PTFE) 0.2μm 100/Pk.
Pack of 100 for $522.00
Add to cart
 
09-924-585 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APUNYL
Nylon (NYL) 0.45μL; 100/Pk.
Pack of 100 for $522.00
Add to cart
 
09-923-252
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
UN203APEPP
Polypropylene (PP) 0.2μm 100/Pk.
Pack of 100 for $522.00
Add to cart
 
Description

Description

Whatman Mini-UniPrep™ Amber Syringeless Filters quickly and easily remove particulates from light-sensitive samples being prepared for HPLC/UHPLC analysis, while meeting USP specifications for light resistance.

  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer

Compliance

Meet USP specifications for light resistance.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Amber
Polypropylene (PP)
0.4mL
Yes
Routine HPLC/UHPLC analysis; Composite assays; Content uniformity; Protein precipitation; Solubility testing; Dissolution testing; Sample filtration; Light sensitive samples; Ion chromatography
Vial with incorporated filtration membrane, plunger and cap/septa
Septa: polytetrafluoroethylene (PTFE) coated silicone rubber
0.6 bar / 8 psi (to compress)
Equivalent in size to 12 x 32mm vials 
50°C
Autosampler, Compressor
Syringeless Filter
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit