Whatman™ Mini-UniPrep™ Amber Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.
Produced with the same colorant used in pharmaceutical containers
Manufacturer: Whatman™ US203APUNYL
 View more versions of this product
Catalog No. 09-800-990
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Pore Size 0.45μm
    Type Syringeless Filter
    Material Nylon
  • Description & Specifications
Related Products
Product Content Feedback