Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman Mini-UniPrep Syringeless Filters
Processes sample loads in one-third the time. Whatman Mini-UniPrep Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
$413.00 - $3485.00
Specifications
Applications | Routine HPLC/UHPLC analysis; Composite assays; Content uniformity; Protein precipitation; Solubility testing; Dissolution testing; Sample filtration; Standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput; Light sen |
---|
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
09-923-104
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
UN203NPEPP |
Polypropylene (PP) | 0.2μm | 100/Pk. |
Pack of 100 for $413.00
|
|
09-923-109
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
UN503NPEPP |
Polypropylene (PP) | 0.2μm | 1000/Pk. |
Pack of 1000 for $3,485.00
|
|
Description
- Pre-assembled all-in-one filtration process; eliminates the need for syringe filter, syringe, vial and septum; reduces costs by up to 40%
- Wide range of membrane choices from 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
- Compatible with most major autosamplers
- Fewer consumables required
- Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
- Can be used either manually or with a compressor unit
- Processes up to 6 samples at one time, further improving sample processing time and reducing the risk of hand stress
- Designed to fit into any autosampler accommodating 12 x 32 mm vials
- OR, septum can be pierced with needle and sample drawn off for manual injection into an analyzer
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Routine HPLC/UHPLC analysis; Composite assays; Content uniformity; Protein precipitation; Solubility testing; Dissolution testing; Sample filtration; Standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput; Light sen |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.