Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters with Preslit Septa

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters with Preslit Septa provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

Supplier:  Cytiva US203NPUAQU

 View more versions of this product

Catalog No. 09-924-588


Add to Cart

Description

Description

Eliminate coring problems associated with repeated automated sampling. Available in standard translucent. Use with standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput.

 

  • For high-throughput automation
  • Slit septum cap enables Mini-UniPrep use with current robotics on HPLC instruments for high throughput automation
  • Durable yet flexible slit septum cap has been specially designed for instruments with sensitive sampling needs
  • Sample evaporation is minimal
  • Increase lab productivity and efficiency by processing samples in one-third the time of conventional syringe filters Consist of a 0.5mL capacity chamber and plunger with filtration membrane at one end and a preattached cap/septum at the other
  • Designed to fit into any autosampler that accommodates 12 × 32mm vials, or pierce septum with needle to draw sample off for manual injection into an analyzer

 

Nylon (NYL)

  • Sample type: Inherently hydrophilic, aqueous/organic samples in 3 to 10 pH range
Polytetrafluoroethylene (PTFE)
  • Sample type: Hydrophobic, for samples >50% organic solvents
Polyvinylidene Difluoride (PVDF)
  • Sample type: Aqueous/≤60% organic solvents-low non-specific protein binding media

 

Do not overfill filter unit or compress the device at an angle before the airtight seal is made. Liquid could escape from air vent hole, resulting in sample loss or personal injury.

Applications:

 

  • Routine HPLC/UHPLC analysis
  • Composite assays
  • Content uniformity
  • Protein precipitation
  • Solubility testing
  • Dissolution testing
  • Sample filtration
  • Standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput
  • Light sensitive samples
  • Ion chromatography

 

Order Info

Filters are also available with a flared cap to enable trouble-free operation with Shimadzu™ autosamplers.

Specifications

Specifications

PVDF
Routine HPLC/UHPLC analysis; Composite assays; Content uniformity; Protein precipitation; Solubility testing; Dissolution testing; Sample filtration; Standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput; Light sen
Syringeless Filter
0.45 μm
100/Pk.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit