Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman Mini-UniPrep Syringeless Filters with Preslit Septa
Processes sample loads in one-third the time. Whatman Mini-UniPrep Syringeless Filters with Preslit Septa provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
$587.00 - $4453.00
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
09-923-110
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPEAQU |
PVDF | 0.2μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-111
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPENYL |
Nylon (NYL) | 0.2μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-112
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPEORG |
Polytetrafluoroethylene (PTFE) | 0.2μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-113
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPEPES |
Polyethersulfone (PES) | 0.2μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-114
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPEPP |
Polypropylene (PP) | 0.2μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-117
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUGMF |
Glass Microfiber (GMF) | 0.45μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-119
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUORG |
Polytetrafluoroethylene (PTFE) | 0.45μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-118
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUNYL |
Nylon (NYL) | 0.45μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-126
|
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPEPP |
PP | 0.2μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
09-923-127
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPUAQU |
PVDF | 0.45μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
09-924-588
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUAQU |
PVDF | 0.45μL | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-133
|
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPUPP |
Polypropylene (PP) | 0.45μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-131
|
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPUORG |
Polytetrafluoroethylene (PTFE) | 0.45μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-128
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPUDPP |
Depth Polypropylene (PP) | 0.45μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-125
|
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPEPES |
Polyethersulfone (PES) | 0.2μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-124
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPEORG |
Polytetrafluoroethylene (PTFE) | 0.2μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-122
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US503NPEAQU |
PVDF | 0.2μm | 1000/Pk. |
Pack of 1000 for $4,453.00
|
|
09-923-121
|
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUPP |
Polypropylene (PP) | 0.45μm | 100/Pk. |
Pack of 100 for $587.00
|
|
09-923-116
![]() |
Cytiva (Formerly GE Healthcare Life Sciences)
US203NPUDPP |
Depth Polypropylene (PP) | 0.45μm | 100/Pk. |
Pack of 100 for $587.00
|
|
Description
Eliminate coring problems associated with repeated automated sampling. Available in standard translucent. Use with standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput.
- For high-throughput automation
- Slit septum cap enables Mini-UniPrep use with current robotics on HPLC instruments for high throughput automation
- Durable yet flexible slit septum cap has been specially designed for instruments with sensitive sampling needs
- Sample evaporation is minimal
- Increase lab productivity and efficiency by processing samples in one-third the time of conventional syringe filters Consist of a 0.5mL capacity chamber and plunger with filtration membrane at one end and a preattached cap/septum at the other
- Designed to fit into any autosampler that accommodates 12 x 32mm vials, or pierce septum with needle to draw sample off for manual injection into an analyzer
Nylon (NYL)
- Sample type: Inherently hydrophilic, aqueous/organic samples in 3 to 10 pH range
- Sample type: Hydrophobic, for samples >50% organic solvents
- Sample type: Aqueous/≤60% organic solvents-low non-specific protein binding media
Do not overfill filter unit or compress the device at an angle before the airtight seal is made. Liquid could escape from air vent hole, resulting in sample loss or personal injury.
Applications:
- Routine HPLC/UHPLC analysis
- Composite assays
- Content uniformity
- Protein precipitation
- Solubility testing
- Dissolution testing
- Sample filtration
- Standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput
- Light sensitive samples
- Ion chromatography
Order Info
Filters are also available with a flared cap to enable trouble-free operation with Shimadzu autosamplers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Syringeless Filter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.