Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters PROMO

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

$156.50 - $1421.00

Specifications

Color Translucent
Housing Material Polypropylene (PP)
Pressure Rating 0.6 bar / 8 psi (to compress)
Volume (Metric) 0.4 mL
Description Equivalent in size to 12 x 32mm vials
View More Specs

Products 3
Catalog Number Mfr. No. Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size No. per Pack Price Quantity & Availability  
09-923-102LR1
View Documents
Cytiva
29648069
0.2 μm 100
Pack of 100 for $156.50
Add to cart
 
09-923-107LR1
View Documents
Cytiva
29648068
0.2 μm 1000
Pack of 1000 for $1,421.00
Add to cart
 
09-923-28LR1
View Documents
Cytiva
29648070
0.45 μm 100
Pack of 100 for $156.50
Add to cart
 
Description

Description

Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.

  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications

Specifications

Translucent
0.6 bar / 8 psi (to compress)
Equivalent in size to 12 x 32mm vials
50°C
Autosampler, Compressor
Syringeless Filter
Polypropylene (PP)
0.4 mL
Yes
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Vial with incorporated filtration membrane, plunger and cap/septa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit