Learn More
GE Healthcare Whatman™ Mini-UniPrep™ Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Manufacturer:  GE Healthcare UN503NPUPP
 View more versions of this product
Catalog No. 09-923-38
Add to cart
Description
Description
  • Pre-assembled all-in-one filtration process; eliminates the need for syringe filter, syringe, vial and septum; reduces costs by up to 40%
  • Wide range of membrane choices from 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Processes up to 6 samples at one time, further improving sample processing time and reducing the risk of hand stress
  • Designed to fit into any autosampler accommodating 12 x 32 mm vials
  • OR, septum can be pierced with needle and sample drawn off for manual injection into an analyzer
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Syringeless Filter
Routine analysis; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Light sensitive samples; Ion chromatography
Polypropylene (PP)
Polypropylene (PP)
0.45μm
1000/Pk.
Documents
Accessories