Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Mini-UniPrep™ Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

Manufacturer:  GE Healthcare UN203NPUPP

 View more versions of this product

Catalog No. 09-923-31


Add to cart

Description

Description

Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.

  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Autosampler, Compressor
Septa: polytetrafluoroethylene (PTFE) coated silicone rubber
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Polypropylene (PP)
0.45μm
Translucent
Equivalent in size to 12 x 32mm vials 
50°C
Syringeless Filter
0.6 bar / 8 psi (to compress)
Yes
0.4mL
Polypropylene (PP)
100/Pk.
Vial with incorporated filtration membrane, plunger and cap/septa
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit