Learn More
GE Healthcare, Whatman Minifold I Spot-Blot System, complete 96-well (acrylic) Available on GSA/VA Contract for Federal Government customers only.
Minifold I Spot-Blot system, complete 96 well (acrylic)
Manufacturer:  GE Healthcare, Whatman 10447850
 View more versions of this product
Catalog No. 09-927-322
  Check Availability
Add to cart
Specifications
Description
  • Easy-to-assemble system.
  • Available in three interchangeable configurations for spot-, dot-, and slot-blots.
  • Accepts all types of transfer membranes: Allows for choice of membrane with the highest binding capacity.
Description & Specifications
Specifications
Minifold I Spot-Blot System
Acrylic 96-well manifold apparatus, 5 x Protran BA85 sheets, 5 x 3mm Chr sheets
Acrylic
200uL/well