Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membrane Filters
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
$204.00 - $1122.00
Specifications
Autoclavable Autoclavable
Material MCE
Sterility Sterile
View More Specs
Products 15
Catalog Number Mfr. No. Pore Size Diameter (Metric) Color Membrane Type Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Diameter (Metric) Color Membrane Type Price Quantity & Availability  
09-927-366
View Documents
Cytiva
10407314
0.45 μm 50 mm White ME25/21 STL
Pack of 400 for $774.00
Add to Cart
09-806-216
View Documents
Cytiva
10406870
0.45 μm 47 mm White ME25/21 ST
Pack of 100 for $204.00
Edge
Add to Cart
09-927-799
View Documents
Cytiva
10406871
0.45 μm 47 mm White ME25
Pack of 1000 for $1,001.00
Add to Cart
09-927-794
View Documents
Cytiva
10406872
0.45 μm 50 mm White ME25/21 ST
Pack of 100 for $262.00
Add to Cart
09-927-363
View Documents
Cytiva
10407332
0.45 μm 47 mm Black ME25/31
Pack of 400 for $767.00
Add to Cart
09-927-724
View Documents
Cytiva
10407334
0.45 μm 50 mm Black ME25/31
Pack of 400 for $774.00
Add to Cart
09-927-778
View Documents
Cytiva
10407370
0.45 μm 47 mm Green ME25/41 STL
Pack of 400 for $755.00
Add to Cart
09-927-719
View Documents
Cytiva
10407372
0.45 μm 50 mm Green ME25/41 STL
Pack of 400 for $781.00
Add to Cart
09-806-218
View Documents
Cytiva
10409470
0.45 μm 47 mm Green ME25/41
Pack of 100 for $263.00
Add to Cart
09-806-219
View Documents
Cytiva
10409472
0.45 μm 50 mm Green ME25/41
Pack of 100 for $265.00
Add to Cart
09-806-214
View Documents
Cytiva
10408712
0.2 μm 47 mm White ME24
Pack of 400 for $847.00
Add to Cart
09-806-215
View Documents
Cytiva
10408714
0.2 μm 50 mm White ME24
Pack of 400 for $760.00
Add to Cart
09-927-795
View Documents
Cytiva
10407324
0.45 μm 50 mm White ME25
Pack of 400 for $787.00
Add to Cart
09-927-217
View Documents
Cytiva
10408915
0.8 μm 50 mm White ME27
Pack of 400 for $1,122.00
Add to Cart
09-927-280
View Documents
Cytiva
10409834
0.6 μm 47 mm Black ME26
Pack of 400 for $839.00
Add to Cart
Description
Description
  • Sterile options available for critical applications
  • Contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
Specifications
Specifications
Autoclavable
MCE
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.