Learn More
GE Healthcare Whatman™ Mixed Cellulose Ester Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
Manufacturer:  GE Healthcare 10407332
 View more versions of this product
Catalog No. 09-927-363
Add to cart
Description
Description
  • Sterile options available for critical applications
  • Contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
1.85 in.
White
Mixed Cellulose Ester, Mixed Cellulose Ester (MCE)
0.45μm, 0.45μm
0.31cm
Grid
130°C
To quantitatively evaluate microorganism counts
47mm
400
Black
Membrane butler
Hydrophilic
ME25
Medium
Sterile, Sterile
266°F
4.7cm
Membrane
Membrane Filter