Whatman™ Nuclepore™ Polycarbonate Track-Etched Membrane, 8 x 10 in. Sheet
Hydrophilic, with low protein binding and low extractables ensuring no sample contamination
Manufacturer: Whatman™ 113516
 View more versions of this product
Catalog No. 09-927-979
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    Specifications
    Pore Size 12.0μm
    Material Polycarbonate
    Quantity (No. per Case) 25/Pk.
    Filter Type Track-Etched Membrane
    Hydrophobicity Hydrophilic
    Manufactured from high-quality polycarbonate film and with sharply defined pores, high flow rates, and excellent chemical and thermal resistance.
    • Smooth flat surface for good visibility of particles
    • Very low level of extractables
    • Chemical resistance and thermal stability support processing a wide variety of samples
    • Low, consistent ash and low tare weights
  • Description & Specifications
Related Products
Product Content Feedback