Whatman™ Nuclepore™ Track-Etched Polycarbonate Gold-Coated Membranes Available on GSA/VA Contract for Federal Government customers only.
Exhibits low protein binding and low extractables ensuring no sample contamination. Whatman™ Nuclepore™ Track-Etched Polycarbonate Gold-Coated Membranes with smooth, flat surface provide excellent visibility of particles.
Manufacturer: Whatman™ 170607
Catalog No. 09-874-624
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Diameter (Metric) Outer 25mm
    Wettability Hydrophilic
    Material Nuclepore Polycarbonate
    Membrane Type Track-etched
    Quantity (No. per Case) 50Pack
    Pore Size 0.4μm
    • Low protein binding and low extractables ensuring no sample contamination.
    • High chemical resistance and good thermal stability for a wide range of samples.
    • Low, consistent ash and low tare weights.
    • Smooth flat surface for good visibility of particles.
    • Nuclepore track-etched polycarbonate membranes are manufactured from high-quality polycarbonate film and have sharply defined pore sizes, high flow rates, and excellent chemical and thermal resistance.
    • The membranes have a smooth flat surface and exhibit very low levels of extractables.
  • Description & Specifications
Related Products
Product Content Feedback