Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Manufacturer: Whatman™ 110809
 View more versions of this product
Catalog No. 09-924-530
  Check Availability
Add to Cart
    Specifications
    Material Nuclepore Polycarbonate
    Wettability Hydrophilic
    Membrane Type Track-etched
    Quantity (No. per Case) 100Pack
    Pore Size 0.8μm
    Diameter (Metric) Outer 37mm
    Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles.
    • Track-etched for highly defined pore size and high pore density
    • Treated with PVP to render it hydrophilic
    • Low extractables, minimizing sample contamination risk
    • High chemical resistance and good thermal stability for a wide range of sample types
    • Low, consistent ash and low tare weights
    • Smooth flat surface for high particle visibility
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
  • Description & Specifications
Related Products
Product Content Feedback