Learn More
GE Healthcare Whatman™ Nytran™ Nylon Membranes
Cast uniformly on both sides of a support matrix, providing excellent symmetry and allowing membranes to lie flat without curling
Manufacturer:  GE Healthcare 10416085
 View more versions of this product
Catalog No. 09-301-149
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Moderately charged—ideal for applications that require a lower charge including Southern and Northern blotting, colony and plaque lifts, as well as dot and slot blots
  • Allows for excellent signal-to-noise ratios
  • Binding capacity: >400µg/cm2
  • Compatible with isotopic and nonisotopic detection
  • Available in 0.2µm and 0.45µm pore size for optimal retention of oligos and larger DNA fragments
Specifications
Specifications
20cm
Suitable for isotopic and nonisotopic detection
>400μg/cm2
0.2μm
20cm
Nylon
Sheet