Learn More
GE Healthcare Whatman™ Partisil™ HPLC Columns
Irregularly shaped media in analytical, semipreparative, and preparative columns
Manufacturer:  GE Healthcare 4223001
 View more versions of this product
Catalog No. 05-713-080
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Compatible with all HPLC and LC/MS instrumentation
  • Neutral pH silica gel improves peak symmetry without the need for mobile phase modifiers
  • Uniform particle sizing minimizes backpressure
  • Efficiencies for Partisil 5µm media—50,000 N/M; Partisil 10µm media—20,000 N/M
Standard Analytical Columns
  • Allow direct scale-up or scale-down to Magnum™ columns
  • Applied with Whatman Compression Screw (WCS) fittings
RAC™ II Second-Generation Rapid-Analysis Columns
  • Analytical columns for faster separations with reduced solvent consumption
  • Special endfitting eliminates unswept corners to maximize resolution and peak symmetry
  • Operate at low backpressure, even at high flowrates, for longer life
  • Connect to LC instrumentation with WCS fittings
Magnum 9™ Columns
    • Semipreparative columns for microgram-to-gram quantities
    • Coned outlet allows high load capacity with minimal peak distortion
    • Compatible with HPLC instruments, allowing you to use the same equipment for both analytical and preparative work
Specifications
Specifications
Yes
250 mm
ODS
4.6 mm
10 µm