Learn More
GE Healthcare Whatman™ pH Indicators Integral Comparison Strips Available on GSA/VA Contract for Federal Government customers only.
Used for rapid determination of pH values
Manufacturer:  GE Healthcare 2629990
Catalog No. 09-876-20
Add to cart
Description
Description
  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100 x 11mm
Strip
pH indicator Test Strip
6 to 8.1
Routine pH testing
Plastic supports, paper segments on each strip, complementary indicator charts
Integral Comparison Strip