Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Polycap™ Disposable Capsules, 36 TF Available on GSA/VA Contract for Federal Government customers only.

Manufactured with rugged PTFE membrane and polypropylene housing. Whatman™ Polycap™ Disposable Capsules, 36 TF filters organic and aggressive solutions, air and gas.

Manufacturer:  GE Healthcare 67113602

 View more versions of this product

Catalog No. 09-743-103


Add to cart

Description

Description

  • Integrity testable
  • Bidirectional flow: certain applications may require orientation
  • Filter Media: Hydrophobic PTFE membrane

Applications:

Filtration of organic solutions, aggressive solutions, air and gas; also can be used as vent.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Polycap TF 36, 0.2 μm, with MNPT inlet and 3/8 in SB outlet (1 pc)
0.2μm
Polypropylene (PP)
GE’s Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.
500cm2
Stepped barb for 6 to 10mm (1/4 to 3/8 in.) tubing
4.1 bar (60 psi)
PTFE
Yes
Filtration of organic solutions, aggressive solutions, air and gas; also can be used as vent.
Male National Pipe Thread
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit