Whatman™ Polycap Disposable Capsules, 75 AS Available on GSA/VA Contract for Federal Government customers only.
Polycap 75 AS filters for filtration of aqueous solutions
Manufacturer: Whatman™ 67057510
 View more versions of this product
Catalog No. 09-744-60
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Pore Size 1μm
    Material (Membrane) Nylon
    Housing Material Polypropylene
    Sterile Yes
    Autoclavable Yes
    For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
    Filtration Area (Metric) 820cm2
    Inlet Style 1/2 in. Stepped Barb
    Outlet Style 1/2 in. Stepped Barb
    • Sterile and nonpyrogenic
    • Integrity testable
    • With inlet vent
    • Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
    • Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
    Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
  • Description & Specifications
Related Products
Product Content Feedback