Whatman™ Polycap Disposable Capsules, 75 AS
Polycap 75 AS filters for filtration of aqueous solutions
Manufacturer: Whatman™ 67067502
 View more versions of this product
Catalog No. 09-744-62
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
    Filtration Area (Metric) 820cm2
    Inlet Style Stepped Barb with filling bell
    Outlet Style Stepped Barb with filling bell
    Pore Size 0.2μm
    Material (Membrane) Nylon
    Housing Material Polypropylene
    Sterile Yes
    Autoclavable Yes
  • Description & Specifications
Related Products
Product Content Feedback