Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Whatman™ Polycap Disposable Capsules, 75 AS
Polycap 75 AS filters for filtration of aqueous solutions
Manufacturer:  Whatman™ 67087502
 View more versions of this product
Catalog No. 09-743-92
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
0.2μm
Polypropylene
Yes
820cm2
1/2 in. Stepped Barb
Nylon
Yes
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
1/2 in. Stepped Barb