Whatman™ Polycap Disposable Capsules, 75 AS
Polycap 75 AS filters for filtration of aqueous solutions
Manufacturer: Whatman™ 67087504
 View more versions of this product
Catalog No. 09-744-100
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
    Filtration Area (Metric) 820cm2
    Inlet Style 1/2 in. Stepped Barb
    Outlet Style 1/2 in. Stepped Barb
    Pore Size 0.45μm
    Material (Membrane) Nylon
    Housing Material Polypropylene
    Sterile Yes
    Autoclavable Yes
  • Description & Specifications
Related Products
Product Content Feedback