Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 75 AS
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Outlet Style:
Female National Pipe Thread
Stepped Barb
Sterility:
Non-sterile
Sterile
Description
- Sterile and nonpyrogenic
- Integrity testable
- With inlet vent
- Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
- Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.

Specifications
Specifications
Autoclavable | Autoclavable |
Description | Polycap AS 75 Capsule Filter, 0.45 μm, SB inlet and outlet (5 pcs) |
Material (Membrane) | Nylon |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. |
Housing Material | Polypropylene |
Inlet Style | Stepped Barb |
Max. Pressure | 4.1 bar (60 psi) |
Outlet Style | Stepped Barb |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction