Learn More
Cytiva Whatman™ Polycap™ HD 36 Disposable Capsule Filters
Ensures a finishing step yielding high purity final product. Whatman™ Polycap™ HD 36 Capsule Filters are suitable for filtering chemical solutions across wide temperature and pH ranges.
$125.00 - $587.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material (Membrane) | Monofilament Anisotropic Polypropylene |
For Use With (Application) | Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. |
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity & Availability | |||
09-744-19
|
Cytiva
67033611 |
10 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
Each for $129.70
|
|
|||
09-744-18
|
Cytiva
67033610 |
1 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
Each for $129.70
|
|
|||
09-744-25
|
Cytiva
67033650 |
5 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
Each for $129.70
|
|
|||
09-744-22
|
Cytiva
67033621 |
20 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
Each for $125.00
|
|
|||
09-927-238
|
Cytiva
2610T |
0.2 μm | FNPT | FNPT |
Pack of 5 for $571.50
|
|
|||
09-927-457
|
Cytiva
2613T |
10 μm | FNPT | FNPT |
Pack of 5 for $587.00
|
|
|||
09-927-462
|
Cytiva
2611T |
1 μm | FNPT | FNPT |
Pack of 5 for $587.00
|
|
|||
09-927-840
|
Cytiva
2612T |
5 μm | FNPT | FNPT |
Pack of 5 for $571.50
|
|
|||
09-927-864
|
Cytiva
2614T |
20 μm | FNPT | FNPT |
Pack of 5 for $571.50
|
|
|||
09-927-402
|
Cytiva
2611 |
1 μm | Stepped Barb 10 to 12mm (3/8 to 1/2 in.) | Stepped Barb 10 to 12mm (3/8 to 1/2 in.) |
Pack of 5 for $587.00
|
|
|||
Description
Ensures a finishing step yielding high purity final product. Whatman™ Polycap™ HD 36 Capsule Filters are suitable for filtering chemical solutions across wide temperature and pH ranges.
Filtration of aqueous solutions, biologicals, inks and pigments, photographic emulsions, food and beverages, solvents. Offer broad capacity between gross paper filters and microporous membranes.- Integrity testable
- With inlet vent
- Filter Media: Monofilament anisotropic polypropylene (MAPP) filter
- Able to be sterilized by autoclaving with steam or ethylene oxide (EtO)
- Retention (with air purge), <2mL
- Max pressure: 4.1 bar (60 psi)
Specifications
Autoclavable | |
Monofilament Anisotropic Polypropylene | |
Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. |