Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polydisc HD Inline Filters

For in-line filtration and isolation of larger volume samples

Supplier:  Cytiva 2228

Catalog No. 09-927-390


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Available options for filtration media suit a range of aqueous and organic samples.

 

  • All polypropylene unit with broad solvent compatibility
  • 5 and 10μm pore size
  • Filter up to one liter with one device
  • High-purity polypropylene housing maintains sample purity
  • Versatile, reusable filters designed for large volume samples
  • Options for inlet and outlet include NPT and Stepped Barb that fits 1/4 to 3/8 in. hoses

 

 

  • Lightweight (11.5g)
  • 50mm filter devices
  • Autoclavable

 

Specifications

Specifications

Polydisc HD Filter, polypropylene, 10.0 μm (50 pcs)
20.4 cm2
NPT
50
50 mm
10 μm
Polypropylene (PP)
50 Pieces
Up to 1000 mL
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit