Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ PolyVENT™ Vessel Venting Filters Available on GSA/VA Contract for Federal Government customers only.

Flows in both directions with hydrophobic PTFE membrane. Whatman™ PolyVENT™ Vessel Venting Filters are integral filter products for sterile venting of vessels and tanks.

Manufacturer:  GE Healthcare 67131651

 View more versions of this product

Catalog No. 09-924-560


Add to cart

Description

Description

Constructed from a single, standardized set of materials.
  • Membrane made of hydrophobic 0.2 µm PTFE membrane.
  • Validated for 50 steam autoclave cycles; compatible with EtO.
  • Testable by Water Break Through (WBT) test or bubble point testing.
  • Pass USP Class VI biosafety tests for plastics.
  • Manufactured in clean room facilities.
  • Bidirectional and autoclavable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

  • Biosafe—all materials meet USP Class VI requirements
  • Manufactured in GMP-controlled cleanroom facilities

Specifications

Specifications

PolyVENT 16, 0.2 μm, inlet 1/4 to 3/8 in. stepped hose barb, outlet 1/4 to 3/8 in. stepped hose barb (100 pcs)
2 bar - forward direction (29 psi - forward direction)
0.2μm
Polypropylene (PP)
100 pieces
Biosafe; all materials meet USP Class VI requirements; Manufactured in GMP-controlled cleanroom facilities
46mm
100 Pieces
Stepped barb 6 to 9mm (¼ to ⅜ in.)
1/4 to 3/8 in SB
53mm
PTFE
Yes
GE’s Whatman PolyVENT filters are integral venting filters that work bidirectionally to prevent contaminants from entering vessels like fermentation tanks during draining or filling.
Sterile venting of filling, mixing, holding, autoclave, fermentation, shipping, and dispensing vessels; For isolation of incubators, autoclaves, lyophilizers, EtO sterilizers, and fermenters; For biotech, pharmaceutical and diagnostics, and food and beverage
16 cm2
Stepped barb 6 to 9mm (¼ to ⅜ in.)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit