Learn More
GE Healthcare Whatman™ Protran™ Blotting Membrane Sandwiches
Consist of Protran transfer membrane between 3MM CHR blotting paper
Manufacturer:  GE Healthcare 10485376
 View more versions of this product
Catalog No. 09-925-102
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Blotting Sandwich Features
  • Available in sizes that match the most commonly used precast gels
  • Precut for speed and convenience
Protran Membrane Features
  • Universal membrane suitable for Western, Southern, and Northern blotting
  • 100% nitrocellulose
  • Very high binding capacity
  • Hydrophilic, so no prewetting with methanol is required
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
Specifications
Specifications
7cm
Protran BA83 Membrane w/3MM Chr Paper
0.2μm
8.5cm
Blotting Sandwich