Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2µm BA83 Protran

100% pure nitrocellulose to ensure the highest binding capacity possible

Manufacturer: Whatman™ 10402493

 View more versions of this product

Catalog No. 09-301-127

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Length (Metric) 6.3cm
    Width (Metric) 22.8cm
    For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
    Material Nitrocellulose
    Binding Capacity High
    Format Sheet
    Pore Size 0.2μm
    Quantity 10/Pk.

    • Universal membrane suitable for Western, Southern, and Northern blotting
    • Hydrophilic, so no prewetting with methanol is required
    • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
    • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
    • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
    • Three pore sizes available—0.45µm is ideal for larger molecular weight samples, 0.2µm ensures binding of samples below 20kDa, 0.1µm is used for molecules below 7kDa
    • Also available in circular formats

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit