Learn More
GE Healthcare Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2µm BA83 Protran
100% pure nitrocellulose to ensure the highest binding capacity possible
Manufacturer:  GE Healthcare 10401396
 View more versions of this product
Catalog No. 09-301-115
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45µm is ideal for larger molecular weight samples, 0.2µm ensures binding of samples below 20kDa, 0.1µm is used for molecules below 7kDa
  • Also available in circular formats
Specifications
Specifications
1 Ea.
3m
Nitrocellulose
Roll
30cm
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
High
0.2μm