Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Puradisc™ Aqua 30mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.

Designed specifically for filtration of environmental samples prior to COD and DOC analysis. Whatman™ Puradisc™ Aqua 30 Syringe Filters minimize carbon leaching from the cellulose acetate membrane that could otherwise lead to sample contamination.

Manufacturer:  GE Healthcare 10462656

 View more versions of this product

Catalog No. 09-302-175


Add to cart

Description

Description

  • Low protein binding, hydrophilic cellulose acetate syringe filter media
  • Prewashed cellulose acetate (CA) syringe filter for reduced organic carbon extractables
  • Designed for sample preparation in support of dissolved organic carbon and chemical oxygen demand analysis
  • Standard inlet and outlet luer connectors
  • Larger filtration area (44% greater in comparison with 25mm), 30mm diameter
  • 0.45 micron Cellulose Acetate membrane
  • Membranes are prewashed to reduce organic carbon levels
  • Designed for aqueous samples

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Syringe Filter
Cellulose Acetate (CA)
White
0.45μm
Male Luer
30mm
For filtration of environmental samples prior to COD and DOC analysis
Polycarbonate
Female Luer Lock
50Pack
Documents
Promotions

Promotions

  • GE Healthcare Life Sciences , Fisherbrand
    Get 5% off your purchase of $500 to $999.99 in GE Healthcare™ Life Sciences products with promo code GESAVE5.
    : GESAVE5

    09-30-2018

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit