Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare, Whatman Puradisc™ FP 13 Polypropylene Syringe Filters, 0.2µm, sterile Available on GSA/VA Contract for Federal Government customers only.

Whatman Puradisc syringe filters from GE Healthcare’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.

Manufacturer:  GE Healthcare, Whatman 10462940

 View more versions of this product

Catalog No. 09-927-409


  Check Availability
Add to cart

Specifications

Description

  • From the Puradisc FP 30 range
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
Description & Specifications

Specifications

13mm
75psi
Yes
Mini-tip
Yes
Polypropylene
<25μL
Female luer lock
0.2μm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit