Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ 30mm Puradisc™ FP Nonsterile Cellulose Nitrate Syringe Filters Available on GSA/VA Contract for Federal Government customers only.

Provide high mechanical strength, high flexibility, high flow rates, and low extractable levels. Whatman™ 30mm Puradisc™ FP Nonsterile Cellulose Nitrate Syringe Filters are an excellent choice for trace element analysis applications.

Manufacturer:  GE Healthcare 10462510

 View more versions of this product

Catalog No. 09-302-173


Add to cart

Description

Description

Cellulose Nitrate syringe well-suited for rapid, routine filtration of samples up to 100m
  • 30mm diameter
  • Larger filtration area (44% greater in comparison with 25mm)
  • Designed for aqueous samples
  • Not suitable for protein recovery applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Non-sterile
30mm
6.9 bar (100 psi)
Polycarbonate (PC)
Up to 100mL
5.7 cm2
Male luer
Up to 100 mL
100 Pieces
30 mm
Cellulose Nitrate (CN)
Black
Whatman Puradisc syringe filters from GE Healthcare’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 mL.
5μm
Female luer lock
Puradisc FP 30 mm Cellulose Nitrate Syringe Filter, 5.0 μm (100 pcs)
26 x 34 mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit