Learn More
Cytiva Whatman™ Puradisc H-PTFE Syringe Filters PROMO
Cytiva Life Sciences Whatman™ Puradisc H-PTFE Syringe Filters can be used for both aqueous and aggressive organic solvents
$71.80 - $142.20
Specifications
Autoclavable Autoclavable
Filter Type Hydrophilic Polytetrafluoroethylene (H-PTFE)
Housing Material Pigment Free Polypropylene
Inlet Style Female Luer Lock
Outlet Style Male Luer
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity & Availability  
09-924-601LR1
View Documents
Cytiva
29648063
13 mm 0.20 μm
Pack of 100 for $142.20
Add to cart
09-924-604LR1
View Documents
Cytiva
29648064
13 mm 0.45 μm
Pack of 100 for $142.20
Add to cart
09-924-595LR1
View Documents
Cytiva
29648065
25 mm 0.20 μm
Pack of 50 for $71.80
Add to cart
09-924-598LR1
View Documents
Cytiva
29648066
25 mm 0.45 μm
Pack of 50 for $71.80
Add to cart
Description
Description
  • Whatman™ Puradisc H-PTFE syringe filter contains a hydrophilic membrane that is chemically stable and inert
  • The H-PTFE membrane exhibits low protein binding and can be used for both aqueous and aggressive organic solvents.
Specifications
Specifications
Autoclavable
Pigment Free Polypropylene
Male Luer
Non-sterile
Hydrophilic Polytetrafluoroethylene (H-PTFE)
Female Luer Lock
5.2 bar (75 psi)