Learn More
Cytiva Whatman™ Pyramid Folded Filter Papers PROMO
Pyramid (square) folded filter paper is easy to use and saves valuable time when undertaking repetitive or multiple analyses.
Manufacturer:  Cytiva 29648047
 View more versions of this product
Catalog No. 09-920-065LR1
Add to cart
Description
Description
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency
Specifications
Specifications
Folded
Filter Paper
210 μm
4
1000 Pk.
Cellulose
110 mm
Nominal Ash Content: 0.06%
92 g/m2