Learn More
Cytiva Whatman™ Pyramid Folded Filter Papers PROMO
Pyramid (square) folded filter paper is easy to use and saves valuable time when undertaking repetitive or multiple analyses.
$296.50 - $390.00
Specifications
Material Cellulose
Quantity 1000 Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Grade Price Quantity & Availability  
Catalog Number Mfr. No. Grade Price Quantity & Availability  
09-920-065LR1
View Documents
Cytiva
29648047
4
Pack of 1000 for $296.50
Add to cart
09-920-063LR1
View Documents
Cytiva
29648049
41
Pack of 1000 for $390.00
Add to cart
Description
Description
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency
Specifications
Specifications
Cellulose
1000 Pk.