Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Pyramid Folded Filter Papers

Easy to use, saves valuable time when undertaking repetitive or multiple analyses

$211.00 - $305.00

Specifications

Diameter (Metric) 125 mm
Format Folded
Material Cellulose
Type Filter Paper
For Use With (Application) Conical Funnels, Repetative or Multiple Analysis
View More Specs

Products 2
Catalog Number Mfr. No. Grade Price Quantity & Availability  
Catalog Number Mfr. No. Grade Price Quantity & Availability  
09-928-109
View Documents Promotion Details
Cytiva
9891128
Grade 6
Pack of 1000 for $211.00
Add to cart
 
09-928-110
View Documents Promotion Details
Cytiva
9892128
Grade 40
Pack of 1000 for $305.00
Add to cart
 
Description

Description

  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that is expected from all Whatman™ Filter Papers
Specifications

Specifications

125 mm
Cellulose
Conical Funnels, Repetative or Multiple Analysis
Folded
Filter Paper
1000/Pk.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit