Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Quadrant Folded Filter Papers

Saves users valuable time and provides ease of use when undertaking repetitive or multiple analyses

$127.80 - $254.50

Specifications

Format Folded
Material Cellulose
Type Filter Paper
For Use With (Application) Conical Filter Funnels in Repetitive or Multiple Analysis
Quantity 500/Pk.
View More Specs

Products 9
Catalog Number Mfr. No. Grade Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Grade Diameter (Metric) Price Quantity & Availability  
09-928-099
View Documents
Cytiva
10380404
Grade 1 110 mm
Pack of 500 for $127.80
Add to cart
 
09-928-100
View Documents
Cytiva
10380405
Grade 1 125 mm
Pack of 500 for $131.10
Add to cart
 
09-928-101
View Documents
Cytiva
10380406
Grade 1 150 mm
Pack of 500 for $148.60
Add to cart
 
09-928-093
View Documents
Cytiva
10380004
Grade 40 110 mm
Pack of 500 for $177.00
Add to cart
 
09-928-094
View Documents
Cytiva
10380005
Grade 40 125 mm
Pack of 500 for $239.50
Add to cart
 
09-928-095
View Documents
Cytiva
10380006
Grade 40 150 mm
Pack of 500 for $254.50
Add to cart
 
09-928-096
View Documents
Cytiva
10380204
Grade 41 110 mm
Pack of 500 for $177.00
Add to cart
 
09-928-097
View Documents
Cytiva
10380205
Grade 41 125 mm
Pack of 500 for $239.50
Add to cart
 
09-928-098
View Documents
Cytiva
10380206
Grade 41 150 mm
Pack of 500 for $254.50
Add to cart
 
Description

Description

  • Saves the time it takes to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analysis
  • Typical applications include: gravimetric analysis for numerous components in cements, clays, iron, and steel products
  • A Primary Filter for separating solid matter from aqueous extracts in general soil analysis, quantitative determination of sediments in milk, and as a pure analytical grade clean-up filter for solutions prior to atomic absorption (AA) spectrometry
  • Use as a high-purity filter in the collection of trace elements and radionuclides from the atmosphere
  • Ashless: 0.7% ash maximum for Grades 4 to 44
  • Suitable for a wide range of critical analytical filtration procedures
Specifications

Specifications

Folded
Filter Paper
500/Pk.
Cellulose
Conical Filter Funnels in Repetitive or Multiple Analysis
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit