Learn More
GE Healthcare Whatman™ Qualitative Filter Papers – Grade 520 B Available on GSA/VA Contract for Federal Government customers only.
Filters particles of 15µm. Whatman™ Qualitative Filter Papers – Grade 520 B with high wet strength and flow rate are designed for technical use.
Manufacturer:  GE Healthcare 10331553
 View more versions of this product
Catalog No. 09-927-215
Add to cart
Description
Description
Qualitative filter papers for technical use
  • Manufactured from strengthened cellulose
  • Flat circle format
  • High wet strength
  • High flow rate
Qualitative filter papers for technical use
  • Manufactured from strengthened cellulose
  • Flat circle format
  • High wet strength
  • High flow rate
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
520b FF 320 mm 50/Pk
320 mm
Grade 520b
20 μm
500μm
Grade 520bll ½ thick, pleated filter paper with high wet strength offering a very high flow rate.
12.5 in.
Cellulose
Circle