Learn More
GE Healthcare Whatman™ Qualitative Filter Papers- Grade 604 1/2 Available on GSA/VA Contract for Federal Government customers only.
The choice for gravimetric analysis
Manufacturer:  GE Healthcare 10312751
 View more versions of this product
Catalog No. 09-927-196
Add to cart
Description
Description
  • High wet strength
  • High-purity hardened filter
  • Chemical-resistant to handle vacuum filtration or acid/alkali solutions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
604 FF 240 mm 100/Pk
240 mm
Grade 604
25 μm
Grade 604½ Qualitative Filter Papers for coarse precipitates.
9.4 in.
Cellulose
Circle