Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Qualitative Filter Papers- Grade 604 1/2

The choice for gravimetric analysis

$67.60 - $130.00

Specifications

Format Circle
Grade Grade 604
View More Specs

Products 5
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09-927-196
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10312751
240 mm
Pack of 100 for $82.10
Add to cart
 
09-927-195
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10312747
185 mm
Pack of 100 for $67.60
Add to cart
 
09-927-194
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10312745
150 mm
Pack of 100 for $68.30
Add to cart
 
09-927-191
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10312744
125 mm
Pack of 100 for $68.30
Add to cart
 
09-927-198
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10312753
320 mm
Pack of 100 for $130.00
Add to cart
 
Description

Description

  • High wet strength
  • High-purity hardened filter
  • Chemical-resistant to handle vacuum filtration or acid/alkali solutions

Specifications

Specifications

Circle
Grade 604
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit