Learn More
Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Circles
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
Supplier: Cytiva 1001150
Description
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids.
- Particle Retention: >11μm
- Porosity: medium
- Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
- Surface: smooth
- Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
- Used for soil analysis and seed testing procedures in the agricultural industry
- Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
- Atmospheric dust is collected from airflow and the stain-intensity measured
- Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection
Specifications
13sec./100mL/sq. in. | |
Grade 1 Qualitative Filter Paper Standard Grade, circle, 150 mm | |
Cellulose | |
150 sec. | |
A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids. | |
Medium | |
180 μm | |
Ø 150 mm | |
150 mm |
87 g/m2 | |
Disc | |
>11 μm | |
57mL/min. (water) | |
Grade 1 | |
100/Pk. | |
Qualitative Filter Paper | |
5.9 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.