Whatman™ Quantitative Filter Papers - Grade 41 Available on GSA/VA Contract for Federal Government customers only.

Contains extremely low ash content at 0.007%. Whatman™ Quantitative Filter Papers – Grade 41 aid analytical procedures involving coarse particles or gelatinous precipitates.

Manufacturer: Whatman™ 14416309

Catalog No. 09-927-642

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Quantitative air pollution analysis
    Pore Size (Metric) 20-25μm
    Thickness 220μm
    Diameter (Metric) 25mm
    Quantity 10000 per pack
    Grade 41
    Format Circle

    Gravimetric filter is well suited for quantitative analysis of soil and air.

    • Fastest ashless filter paper
    • For quantitative air pollution analysis
    • Easily removed for further analysis or culturing
    • Disposable filter funnel available

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit