Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Reeve Angel 201 Smooth Qualitative Filter Paper: Circles

For routine, noncritical laboratory filtration applications

Manufacturer:  GE Healthcare 5201240

 View more versions of this product

Catalog No. 09-805-13


Add to cart

Description

Description

  • Retentivity: medium-fine (7-14µm)
  • Filtration speed: slow-medium
  • Color: White

Specifications

Specifications

100/Pk.
201
Slow Medium
24cm
Medium Fine
Circle
Documents
Promotions

Promotions

  • GE Healthcare Life Sciences , Fisherbrand
    Get 5% off your purchase of $500 to $999.99 in GE Healthcare™ Life Sciences products with promo code GESAVE5.
    : GESAVE5

    09-30-2018

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit