Learn More
Whatman™ Reeve Angel Qualitative Filter Papers - Grade 305
For large particles and gelatinous precipitates
Manufacturer:  Whatman™ 5305125
 View more versions of this product
Catalog No. 09-927-736
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
12.5cm
305
20 to 25μm
Circle
5 in.
Cellulose
Fast